Learn More
Invitrogen™ Goat anti-Canine IgA Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca:  Invitrogen™ SA510251
Codice prodotto. 17139635
  • 124.00€ / 1mg
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
This antibody may cross react with IgA from other species.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Canine IgA
Polyclonal
Unconjugated
Goat
Antigen affinity chromatography
RUO
Canine
Secondary Antibody
IgG
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Canine IgA.
1 mg
Secondary
4°C
Liquid
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni