missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Canine IgG Fc Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ SA510272
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody may cross react with IgG from other species.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Canine IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Canine | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Canine IgG Fc. | |
| 1 mg | |
| Secondary | |
| 4°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto