missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Donkey IgG (H+L) Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ PA130377
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PA1-30377 detects Donkey IgG from donkey samples.
Anti-Donkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for equine (Donkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Donkey IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Donkey | |
| Secondary Antibody | |
| IgG |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| 1.0 mg/mL | |
| TBS with 0.001M MgCl2, 0.0001M ZnCl2, 50% glycerol, 1% BSA and 0.01% sodium azide; pH 8 | |
| Donkey IgG whole molecule. | |
| 1 mg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto