Learn More
Goat anti-Ferret IgA/IgG/IgM (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP1-73396
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Ferret samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifica
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Biotin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| This product is suitable for the detection of all Ferret immunoglobulin classes, isotypes and chain combinations. | |
| Ferret | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Ferret IgG, IgA and IgM whole molecules | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Titolo del prodotto
For Research Use Only