Learn More
HADH, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HADH.
Marca: Abnova PAB7412.100ug
Descrizione
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene. [provided by RefSeq
Sequence: C-YERGDASKEDIDSpecifica
| HADH | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| HADH | |
| HADH | |
| A synthetic peptide corresponding to human HADH. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of HADH. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HAD/HADH1/HADHSC/HHF4/M/SCHAD/MGC8392/SCHAD | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3033 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only