missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgE Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ SA510259
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody may cross react with IgE from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Human IgE | |
| Polyclonal | |
| Unconjugated | |
| IGHA1 | |
| HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; Km | |
| Human IgE. | |
| 1 mg | |
| Secondary | |
| Human | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01834, P01857, P01859, P01860, P01861, P01876, P01877, P0CG04 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3493, 3494, 3500, 3501, 3502, 3503, 3514, 3537 | |
| 4°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto