Learn More
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, Rhodamine
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ 31656
Descrizione
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31656 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31656 reacts with the heavy chains of human IgG and with the light chains common to most human immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Rhodamine Amax= 550 nm; Emax= 570 nm. Fluorophore/Protein Absorbance Ratio: A550/A280 = ~ 0.4 (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Human IgG (H+L) | |
| Polyclonal | |
| Rhodamine | |
| PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
| Affinity chromatography | |
| RUO | |
| Secondary | |
| 1.5mL of distilled water (1.5mg/mL after restoration) | |
| Human | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| 1.5 mg/mL | |
| Goat anti-Human IgG (H+L) Secondary Antibody, 2mg, 1.5mg/mL Concentration, Liquid, Rhodamine Conjugate, Goat Host Species, Human Species Reactivity, PBS, pH 7.6 with 15mg/mL BSA Storage Buffer, 4°C Storage Requirements, For IF, ICC, IHC, Flow and IP | |
| Goat | |
| 2 mg | |
| Whole antibody | |
| 1 Each | |
| 0.05% Sodium azide | |
| 4°C | |
| Lyophilized | |
| HRP, AP, fluorescence |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.