missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgA/IgG/IgM (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP2-69275
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
Specifica
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.2) | |
| Highly purified IgG and pools of homogenous IgA and IgM isolated from monkey serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 10 mg | |
| Secondary | |
| Monkey | |
| Lyophilized |
| Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot | |
| LYOPH | |
| Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto