Learn More
Invitrogen™ Goat anti-Monkey IgG (H+L) Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ PA184629
Descrizione
This antibody has been shown to react specifically with monkey IgG heavy chains and with light chains common to other monkey immunoglobulins by immunoelectrophoresis and ELISA. It reacts with African green monkey, Cynomolgus monkey, Baboon, Chimpanzee, Rhesus Monkey and Human IgG. Goat anti Monkey IgG (H/L) antibody recognizes monkey IgG (heavy and light chains).
Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Monkey IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Monkey | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunodiffusion | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Purified monkey IgG. | |
| 1 mg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.