Learn More
Goat anti-Mouse IgG1 Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Marca:  Novus Biologicals NBP2-69522
Codice prodotto. 18616267
  • 1075.00€ / 1mL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
IgG1 Polyclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifica
Specifica
IgG1
Polyclonal
FITC
Lyophilized from PBS (pH 7.2)
Goat
Ion exchange chromatography
RUO
Secondary
Mouse
Lyophilized
ELISA, Immunofluorescence, Immunohistochemistry (Frozen)
LYOPH
ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen
IgG1, Igh-4, RGD1359539, VH7183
Pools of highly purified homogenous IgG1 isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Immunology
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni