Learn More
Goat anti-Mouse IgM Fc Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP2-69733
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Fc Polyclonal antibody specifically detects IgM Fc in Mouse samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
Specifica
| IgM Fc | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Highly purified homogenous IgM isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunoprecipitation, Immunoelectrophoresis | |
| LYOPH | |
| ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis | |
| Goat | |
| Whole Antiserum | |
| RUO | |
| Secondary | |
| Mouse | |
| Lyophilized |