Learn More
NQO1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NQO1.
Marca: Abnova PAB19680.100ug
Descrizione
This gene is a member of the NAD(P)H dehydrogenase (quinone) family and encodes a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. This protein's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Mutations in this gene have been associated with tardive dyskinesia (TD), an increased risk of hematotoxicity after exposure to benzene, and susceptibility to various forms of cancer. Altered expression of this protein has been seen in many tumors and is also associated with Alzheimer's disease (AD). Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: C-DKGPFRSKKAVLSSpecifica
| NQO1 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of NQO1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DHQU/DIA4/DTD/NMOR1/NMORI/QR1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1728 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NQO1 | |
| NQO1 | |
| A synthetic peptide corresponding to internal region of human NQO1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize reported isoform a (NP_000894.1) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only