Learn More
Goat anti-Porcine IgA Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB724
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifica
| IgA Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Immunoglobulin A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3493 | |
| Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000- 1:30000, ELISA 1:1000-1:30000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000, Immunohistochemistry-Paraffin 1:200-1:2000 | |
| IGHA1 | |
| Pig IgA | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with pig IgA. This may cross react with IgA from other species. | |
| Store at 4C. Do not freeze. |
Titolo del prodotto
For Research Use Only