Learn More
Invitrogen™ Goat anti-Rabbit IgG F(ab')2 Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca:  Invitrogen™ 31234
Codice prodotto. 11895925
  • 156.00€ / 2mg
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31234 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31234 reacts with the light chains of rabbit IgG and with those common to other rabbit immunoglobulins, but does not react with the Fc portion of rabbit IgG. The antibody does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Rabbit IgG F(ab')2
Polyclonal
Unconjugated
Goat
2 mg
Secondary
4°C
Liquid
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
2.4 mg/mL
PBS with no preservative; pH 7.6
Antigen affinity chromatography
RUO
Rabbit
Secondary Antibody
IgG
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni