missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgM Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB7174
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifica
| IgM Heavy Chain | |
| Polyclonal | |
| HRP | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Rabbit IgM | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with rabbit IgM. This may cross react with IgM from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:20000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Rabbit | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto