missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgA Heavy Chain Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP2-68527
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| FITC | |
| PBS | |
| Human IgA | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| 0.5 mg/mL | |
| Flow Cytometry Neat - 1:10, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence Neat - 1:10 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction