missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgG F(ab')2 Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP1-51900
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifica
| IgG F(ab')2 | |
| Polyclonal | |
| Unconjugated | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with F(ab′)2 fragments of human IgG. Cross reactivity with IgA and IgM is negligible. No antibody was detected against non-immunoglobulin serum proteins. Antiserum was cross adsorbed using mouse and rat immunosorbents to remove cross reactive antibodies. Less than 1% cross reactivity to mouse and rat IgG was detected. This antibody may cross react with F(ab′)2 fragments of IgG from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000 | |
| IGHG1 | |
| Human IgG-F(ab')2 Fragment | |
| 1 mL | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| 3500 | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto