Learn More
Goat F(ab')2 anti-Human IgG Heavy Chain Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NBP2-68544
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifica
| IgG Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG | |
| 0.5 mg | |
| Immunology | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 0.5 mg/mL | |
| Borate buffered saline | |
| Goat | |
| Chromatography | |
| RUO | |
| Secondary | |
| Store at 4°C. | |
| IgG |