missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG VisUCyte HRP Polymer, R&D Systems™

Donkey Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems VC004-050
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.
Specifica
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat IgG | |
| 50 mL (2 x 25 mL) | |
| Secondary | |
| Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC. | |
| IgG |
| Immunohistochemistry | |
| HRP Polymer | |
| Supplied as a solution in PBS with preservative. | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto