Learn More
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Codice prodotto. 10371335 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Diametro (sistema metrico) interno:
0.8 mm
1.2 mm
Da utilizzare con (apparecchiatura):
Dual Column Installation
Single Column Installation
Single Column Installation (Cross Version)
Quantità:
10 Pack
2 Pack
Product Type:
Gold Inlet Base Seal
Siltek Inlet Base Seal
Dimensione della confezione:
pezzi 10
pezzi 2
Codice prodotto. 10371335
Marca: Thermo Scientific™ 290GA084
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
TRUSTED_SUSTAINABILITY
Specifica
Quantità 10 Pack
Product Type Gold Inlet Base Seal
Diametro (sistema metrico) interno 0.8 mm
Da utilizzare con (apparecchiatura) Single Column Installation (Cross Version)