Learn More
WTW™ Gold Reagent Test Kit
Click to view available options
N. di test:
80
Dimensione della confezione:
Pezzo
Descrizione
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifica
Specifica
| Da utilizzare con (applicazione) | For waste water, sea water |
| Da utilizzare con (apparecchiatura) | For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Include | Certificates |
| N. di test | 80 |
| Range | 0.5 - 12.0mg/I Au |
| Sufficiente per | 100 Digestions |
| Metodo test | Photometric |
| Tipo | Crack set |