Learn More
GPR116 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14047-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR116 (G protein-coupled receptor 116) is a 1,346 amino acid multi-pass membrane protein that contains one SEA domain, one GPS domain and three Ig-like domains and belongs to the GPR family. Existing as a dilsulfide-liked homodimer at the cell surface, GPR116 exists as multiple alternatively spliced isoforms and is thought to play a role in regulating and maintaining proper acid-base balance throughout the cell.Specifica
| GPR116 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF5 | |
| G protein coupled receptor 116, GPR116, KIAA0758, KPG_001 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221395, 224792, 245977 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| G5E8Q8, Q8IZF2, Q9WVT0 | |
| ADGRF5 | |
| GPR116 Fusion Protein Ag5214 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |