missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA533714
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Rabbit (89%).
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifica
| GPR40 | |
| Polyclonal | |
| Unconjugated | |
| FFAR1 | |
| FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O14842 | |
| FFAR1 | |
| Synthetic 18 amino acid peptide from C-terminus of human GPR40. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto