Learn More
Fisherbrand™ Grade 8 Microglass Fiber Extraction Thimble
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Lunghezza (sistema metrico):
80 mm
Diameter (Metric) Inside:
25 mm
Dimensione della confezione:
pezzi 25
Descrizione
Applications:
- Used in extraction of sulfur in gaseous masses
- Helps in determinimg content of free lipids in food by extraction
- Determines dust and tar in industrial air and gases
- Lacquer and binder analysis in paint colours
- Used in control of emission smokes in industrial chimneys and gravimetric analysis of dust in gases
Specifica
Specifica
| Diameter (Metric) Inside | 25 mm |
| Materiale | Microglass Fiber |
| Da utilizzare con (applicazione) | Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases |
| Grado | 8 |
| Lunghezza (sistema metrico) | 80 mm |
| Temperatura max (sistema metrico) | 500°C |
| Product Type | Extraction Thimble |