Learn More
Ansell™ Comfort G26G Nitrile Immersion Gloves
Click to view available options
Size:
6.5
7.5
8.5
10.5
Dimensione della confezione:
12 paia
Descrizione
- Very good abrasion resistance.
- Allows contact up to 100°C.
- Superior raised grip pattern for safety handlings.
- Open cuff with textile edge binding allows ventilation to keep hand cool.
Specifica
Specifica
| Materiale parte posteriore | Nitrile |
| Materiale polso | Nitrile |
| Product Type | Immersion Gloves |
| Size | 8.5 |
| Colore polso guanto | Verde |
| Stile polso guanto | Guanto con manichetta |
| Materiale rivestimento interno | Cotone |
| Modello | COMFORT G26G |
| Quantità | 12 paia |
| Resistente a | Excellent resistance to many solvents and oil based chemicals |