missing translation for 'onlineSavingsMsg'
Learn More

GSK 4716, Tocris Bioscience™

Catalog Number 18025092
Click to view available options
Quantità:
10 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
101574-65-6
C17H18N2O2
282.34
MFCD00567155
IKPPIUNQWSRCOZ-WOJGMQOQSA-N
e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
5331325
CHEBI:80000
4-hydroxy-N'-[(E)-[4-(propan-2-yl)phenyl]methylidene]benzohydrazide
CC(C)C1=CC=C(\C=N\NC(=O)C2=CC=C(O)C=C2)C=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
101574-65-6
C17H18N2O2
282.34
MFCD00567155
IKPPIUNQWSRCOZ-WOJGMQOQSA-N
e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
5331325
CHEBI:80000
4-hydroxy-N'-[(E)-[4-(propan-2-yl)phenyl]methylidene]benzohydrazide
CC(C)C1=CC=C(\C=N\NC(=O)C2=CC=C(O)C=C2)C=C1

Selective ERRγ and ERRβ agonist

Selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays selectivity over ERRα and the classical estrogen receptors.
Materiale o nome chimico GSK 4716
Quantità 50 mg
Peso formulazione 282.34
Percent Purity >99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'