missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSPT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA560824
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: TQPPTLPAGS GSNDETCTGA GYPQGKRMGR GAPVEPSREE PLVSLEGSNS AVT Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 64%.
ERF3B encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.
Specifica
| GSPT2 | |
| Polyclonal | |
| Unconjugated | |
| GSPT2 | |
| eRF3b; Eukaryotic peptide chain release factor GTP-binding subunit ERF3B; eukaryotic peptide chain release factor subunit 3b; FLJ10441; G1 to S phase transition 2; G1 to S phase transition protein 2 homolog; GSPT2; GST2; RGD1563213 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23708 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IYD1 | |
| GSPT2 | |
| Recombinant protein corresponding to Human GSPT2. Recombinant protein control fragment (Product #RP-100487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto