Learn More
Cytiva GST Gene Fusion System pGEX Vectors
Codice prodotto. 10534435 Sfoglia Tutto Cytiva Prodotti
Click to view available options
Contenuto kit:
2T
4T-1
4T-3
6P-1
6P-2
6P-3
Dimensione della confezione:
25µL
25µg
Codice prodotto. 10534435
Marca: Cytiva 28954549
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
  • Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
  • Offers an internal laq1q gene for use in any E. coli host
  • With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
  • The vectors provide all three translational reading frames beginning with the EcoRI restriction site
  • Can use a wide variety of E. coli host strains cloning and expression
  • pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
  • pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [γ-32P]ATP using protein kinase
TRUSTED_SUSTAINABILITY
Specifica
Contenuto kit 4T-1
Quantità 25 μg
Da utilizzare con (applicazione) Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
Tipo pGEX Vector