Learn More
Checkers Guard Dog™ Low Profile
Codice prodotto. 17782408
Click to view available options
Lunghezza (sistema metrico):
27 cm
43 cm
72 cm
77 cm
84 cm
Dimensione della confezione:
Pezzo
Codice prodotto. 17782408
Marca: Checkers PGDS0008OB
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market. These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
  • Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market
  • These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
Specifica
Lunghezza (sistema imperiale) 33.1 in.
Lunghezza (sistema metrico) 84 cm
Larghezza (sistema metrico) 91 cm
Colore Black, Orange
Materiale Polyurethane
Tipo Cable Protector