missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY2F Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25252-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GUCY2F (Guanylate cyclase 2F, retinal) is also named as GUC2F, RETGC2 and belongs to the adenylyl cyclase class-4/guanylyl cyclase family. GUCY2F may play a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction. By in situ hybridization, mRNA encoding GUCY2F is found only in the outer nuclear layer and inner segments of photoreceptor cells. By using synthetic peptide antiserum specific for each RetGC subtype, RetGC-2(GUCY2F) can be distinguished from RetGC-1 as a slightly smaller protein in immunoblots of bovine rod outer segments.Specifica
| GUCY2F | |
| Polyclonal | |
| Unconjugated | |
| GUCY2F | |
| CYGF, GC F, guanylate cyclase 2F, retinal, Guanylate cyclase F, GUC2DL, GUC2F, GUCY2F, RETGC 2, RETGC2, Retinal guanylyl cyclase 2, ROS GC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245650, 2986 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P51841, Q5SDA5 | |
| Gucy2f | |
| GUCY2F Fusion Protein Ag19266 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto