missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADH Polyclonal Antibody

HADH Polyclonal Antibody
Marca: Invitrogen PA518840
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HADH Polyclonal Antibody for Western Blot
Specifica
| HADH | |
| Polyclonal | |
| Unconjugated | |
| HADH | |
| AA409008; AU019341; AW742602; Had; HADH; HADH1; HADHSC; HCDH; HHF4; hydroxyacyl-CoA dehydrogenase; hydroxyacyl-Coenzyme A dehydrogenase; hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; hydroxylacyl-Coenzyme A dehydrogenase short chain; hydroxylacyl-Coenzyme A dehydrogenase, short chain; hydroxylacyl-Coenzyme A dehydrogenase-dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; M/SCHAD; medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase; medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; MGC8392; Mschad; SCHAD; short chain 3-hydroxyacyl-CoA dehydrogenase; short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3033 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q16836 | |
| HADH | |
| Synthetic peptide sequence (YERGDASKEDID) corresponding to the internal amino acids of HADH (aa 234-246). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto