missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HADH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA586005
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HADH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.
Specifica
| HADH | |
| Polyclonal | |
| Unconjugated | |
| HADH | |
| AA409008; AU019341; AW742602; Had; HADH; HADH1; HADHSC; HCDH; HHF4; hydroxyacyl-CoA dehydrogenase; hydroxyacyl-Coenzyme A dehydrogenase; hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; hydroxylacyl-Coenzyme A dehydrogenase short chain; hydroxylacyl-Coenzyme A dehydrogenase, short chain; hydroxylacyl-Coenzyme A dehydrogenase-dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; M/SCHAD; medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase; medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; MGC8392; Mschad; SCHAD; short chain 3-hydroxyacyl-CoA dehydrogenase; short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113965, 15107, 3033 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q16836, Q61425, Q9WVK7 | |
| HADH | |
| Synthetic peptide corresponding to Human HADH aa 275-314. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto