Learn More
Invitrogen™ HADH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA588030
Descrizione
Immunogen sequence: MAFVTRQFMR SVSSSSTASA SAKKIIVKHV TVIGGGLMGA GIAQVAAATG HTVVLVDQTE DILAKSKKGI EESLRKVAKK KFAENPKAGD EFVEKTLSTI ATSTDAASVV HSTDLVVEAI VENLKVKNEL FKRLDKFAAE HTIFASNTSS LQITSIANAT TRQDRFAGLH FFNPVPVMKL VEVIKTPMTS QKTFESLVDF SKALGKHPVS CKDTPGFIVN RLLVPYLMEA IRLYERGDAS KEDIDTAMKL GAGYPMGPFE LLDYVGLDTT KFIVDGWHEM DAENPLHQPS PSLNKLVAEN KFGKKTGEGF YKYK; Positive Samples: HepG2, Mouse kidney, Mouse pancreas, Rat kidney, Rat liver; Cellular Location: Mitochondrion matrix.
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.
Specifica
| HADH | |
| Polyclonal | |
| Unconjugated | |
| HADH | |
| AA409008; AU019341; AW742602; Had; HADH; HADH1; HADHSC; HCDH; HHF4; hydroxyacyl-CoA dehydrogenase; hydroxyacyl-Coenzyme A dehydrogenase; hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; hydroxylacyl-Coenzyme A dehydrogenase short chain; hydroxylacyl-Coenzyme A dehydrogenase, short chain; hydroxylacyl-Coenzyme A dehydrogenase-dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; M/SCHAD; medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase; medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; MGC8392; Mschad; SCHAD; short chain 3-hydroxyacyl-CoA dehydrogenase; short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113965, 15107, 3033 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16836, Q61425, Q9WVK7 | |
| HADH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-314 of human HADH (NP_0053183). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.