Learn More
Hamilton™ Siringhe Gastight™ serie 1000: Siringhe Luer-Lock, terminale TLL
Codice prodotto. p-186083
Click to view available options
Volume (sistema metrico):
1 mL
1.0 ml
2.5 ml
5 mL
5.0 ml
10 mL
10.0 ml
25 mL
25.0 ml
50 mL
100 mL
Modello:
1001
1002
1005
1010
1025
1050
1100
Tipo di collegamento:
Attacco Luer
Luer Lock
Luer Lock with Slots
Luer-Lock PTFE
Dimensione della confezione:
Pezzo
Codice prodotto. 10611353
Marca: Hamilton™ 85027
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors
  • The TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations
  • The version with slots is used during the attachment to some instruments but does not effect the connection to needles
  • Autoclavable when disassembled, except on 25mL and greater syringes; repeated autoclaving will shorten syringe life
  • Needle point styles 4 and AS are available on request)
Specifica
Materiale PTFE (Tip), PTFE-tipped (Plunger)
Diametro (sistema imperiale) interno 1.28 in.
Diametro (sistema imperiale) esterno 1.45 in.
Diametro (sistema metrico) interno 32.6 mm
Diametro (sistema metrico) esterno 36.9 mm
Da utilizzare con (applicazione) Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Product Type Gastight Syringe
Pressione max 100 psi
Modello 1050
Numero dei punti dell'ago No Needle Included
Quantità Ea.
Volume (sistema metrico) 50 mL
Tipo di collegamento Luer Lock with Slots