Learn More
Hamilton™ 1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Codice prodotto. 10787954
Click to view available options
Volume (sistema metrico):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Pressione max:
100 psi
200 psi
Dimensione della confezione:
Pezzo
Codice prodotto. 10787954
Marca: Hamilton™ 81656
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Syringes are available in sizes ranging from 1mL to 100mL. An easy-to-use twist valve and a positive rear stop on SampleLock prevents loss of sample and plunger blowout. Optional male luer and female luer lock adapter threads on the SampleLock valve making the syringes compatible with a multitude of connectors and fittings. Typical applications are:
  • Headspace application
  • Environmental sample collection and storage
  • Pre-pressurization of gaseous sample for GC analysis
  • Injection of internal standards through the valve port into samples
  • Sample spiking
Sample Lock 1700 and 1000 Series Syringe Terminations
  • Sample Lock (SL) - The sample lock incorporates an On/Off syringe valve with RN needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking. Not autoclavable.
  • Needle Point Styles
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26-22
    Specifica
    Autoclavabile Not Autoclavable
    Materiale Borosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
    Calibro dell'ago 22 G
    Diametro (sistema imperiale) interno 0.57 in.
    Diametro (sistema imperiale) interno ago 0.01 in.
    Diametro (sistema metrico) interno ago 0.41 mm
    Diametro (sistema imperiale) ago esterno 0.02 in.
    Diametro (sistema imperiale) esterno 0.69 in.
    Diametro (sistema metrico) interno 14.6 mm
    Diametro (sistema metrico) esterno 17.7 mm
    Diametro (sistema metrico) ago esterno 0.72 mm
    Da utilizzare con (applicazione) To collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination
    Lunghezza (sistema imperiale) ago 2 in.
    Product Type General Purpose Syringe
    Pressione max 200 psi
    Temperatura max (sistema imperiale) 239°F
    Temperatura max (sistema metrico) 115°C
    Min. Temperature (English) 50°F
    Numero dei punti dell'ago 2
    Min. Temperature (Metric) 10°C
    Quantità Ea.
    Volume (sistema metrico) 10 mL
    Stile del punto dell'ago Sharp, Bevel, Curved, Non-coring
    Tipo di collegamento Sample Lock, Removable