Learn More
Hamilton™ 700 Series Microliter™ 8100/8200 Autosampler Syringe
Click to view available options
Volume (sistema metrico):
10 μL
Dimensione della confezione:
Pezzo
Descrizione
Microliter syringe with a stainless steel plunger that is hand-fitted to the glass barrel and is designed for liquid sampling. Syringe has a removable needle (RN) termination. Available in 10μL volume.
Syringe Terminations
- Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
- Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauge: 26s
Specifica
Specifica
| Materiale | Borosilicate Glass (Barrel), Stainless Steel (Plunger) |
| Calibro dell'ago | 26s G |
| Diametro (sistema imperiale) interno ago | 0.005 in. |
| Diametro (sistema metrico) interno ago | 0.13 mm |
| Diametro (sistema imperiale) ago esterno | 0.019 in. |
| Diametro (sistema metrico) ago esterno | 0.483 mm |
| Lunghezza (sistema imperiale) | 2 in. |
| Lunghezza (sistema imperiale) ago | 1.69 in. |
| Lunghezza (sistema metrico) | 51 mm |
| Product Type | Microliter Syringe |
| Vedi altri risultati |