Learn More
Hamilton™ Chemically Inert Manual Valves
Click to view available options
Tipo:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
Dimensione della confezione:
Pezzo
Descrizione
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifica
Specifica
| Da utilizzare con (applicazione) | For low pressure applications, from a basic on/off flow to a four-way distribution system |
| Materiale | CTFE, PTFE |
| Tipo | 3-3 Manual Valve |