Learn More
Hamilton™ Metal Hub Side Port Needles (Luer Lock)
Click to view available options
Calibro dell'ago:
18 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
Dimensione della confezione:
pezzi 6
Descrizione
Metal Hub Compatible Syringe Terminations
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
Six needles per pack.
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Materiale | Nickel-plated Brass (Hub), Stainless Steel |
| Calibro dell'ago | 22s G |
| Diametro (sistema imperiale) interno | 0.0059 in. |
| Diametro (sistema imperiale) esterno | 0.028 in. |
| Diametro (sistema metrico) interno | 0.15 mm |
| Diametro (sistema metrico) esterno | 0.72 mm |
| Lunghezza (sistema imperiale) ago | 2 in. |
| Product Type | Replacement Needle |
| Stile del punto dell'ago | Cone with Side Port |
| Vedi altri risultati |