Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - Syringes
Click to view available options
Include:
PTFE plunger tip
Plunger tip
Modello:
1002.5
1705.5 BFP
Volume (sistema metrico):
1.25 μL
25 μL
Dimensione della confezione:
Pezzo
Descrizione
Model 1705.5
- Bubble Free Prime
- Expels air from the syringe and valve thanks to an innovative plunger tip
- 1.25mL
- Model 1002.5
- Gastight
- Ideal for dispensing both liquids and gases
- Has a precision-machined PTFE plunger tip which creates a leak-free seal
- 25μL
Specifica
Specifica
| Modello | 1002.5 |
| Volume (sistema metrico) | 1.25 μL |
| Da utilizzare con (apparecchiatura) | Microlab™ 300 pipetteting system |
| Include | PTFE plunger tip |