Learn More
VITLAB™ Misurino in HDPE
Codice prodotto. 10106550
Click to view available options
Capacità (sistema metrico):
15 mL
65 mL
110 mL
350 ml
Dimensione della confezione:
Pezzo
pezzi 12
pezzi 6
Codice prodotto. 10106550
Marca: VITLAB™ 40093
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ HDPE Measuring Scoops are lighter than metal, don’t dent and feature good chemical resistance.
  • Conical in shape with tapered filling edge
Specifica
Capacità (sistema metrico) 15 mL
Colore Natural
Product Type Measuring Scoop
Da utilizzare con (applicazione) Suitable as weighing
Materiale HDPE
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.