Learn More
Ansell HyFlex™ 70-225 Medium Duty String Knit Gloves with Aramid Fiber
Click to view available options
:
9
Descrizione
Aramid fiber medium duty industrial gloves offers high cut-resistance to weight ratio. Ideal for automotive or machinery and equipment industries.
- Five times the cut resistance of leather
- Heat protection
- Hand-to-hand reversible
Specifica
Specifica
| Coating Material | Neptune |
| Colore | Yellow |
| Cut Resistance Rating | ANSI A3 |
| Hand | Ambidestri |
| Latex Free | Yes |
| Materiale | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | 9 |
| Stile polso guanto | Knit |