missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HepaRG™ Serum-free Induction Medium Supplement
Click to view available options
Quantità:
2 mL
Dimensione della confezione:
Pezzo
Descrizione
HepaRG™ Serum-free Induction Medium Supplement is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Serum-free Induction Medium Supplement is specifically used for induction of metabolic activity in HepaRG™ cells in a serum free environment.
Easy media preparation, just thaw, mix, and use
Reproducible, and biologically relevant results with HepaRG™ cells
Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Serum-free Induction Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.
Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).
HepaRG™ is a trademark of BioPredic International.
Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Serum-free Induction Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.
Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).
HepaRG™ is a trademark of BioPredic International.
Specifica
Specifica
| Content And Storage | Supplement should be stored at -20°C |
| Tipo di cellula | HepaRG Cells |
| Classificazione | Serum-free |
| Forma | Frozen |
| Product Type | Hepatocyte Supplement |
| Serum Level | Serum-Free |
| Sterilità | Sterile |
| Senza additivi | No Glutamine |
| Da utilizzare con (applicazione) | Drug Metabolism |
| Linea di prodotti | Gibco |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto