Learn More
Gilson™ HF2 Pump Head for Gilson™ Peristaltic Pump
Codice prodotto. 12316122 Sfoglia Tutto Gilson Prodotti
Click to view available options
Portata:
1 to 340 mL/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 12316122
Marca: Gilson™ F110707
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
  • R1—Single channel pump head
  • R4—Four channel pump head
  • R8—Eight channel pump head
  • R2—Two channel pump head
  • R2HF—Two channel high flow pump head
  • R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
  • MF1—Pump head with one cartridge
  • MF4—Pump head with four cartridges
  • HF2—Pump head with two high flow cartridges
TRUSTED_SUSTAINABILITY
Specifica
Portata 1 to 340 mL/min.