missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Antibody (SBF5 C1.7), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-2696C
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HGF Monoclonal specifically detects HGF in Human samples. It is validated for Western Blot, ELISA.
Specifica
| HGF | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HGF | |
| Recombinant Human HGF | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3082 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| SBF5 C1.7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| SBF5 C1.7 does not cross react with other members of the Skingle-serine protease superfamily such as HGF1/MSP, plasminogen/plasmin. t-PA and u-PA. | |
| Store at 4C in the dark. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto