missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody (H1alpha67), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 891 publication
317.00€ - 771.00€
Specifica
| Diluizione | Western Blot 1:500, Simple Western, Chromatin Immunoprecipitation 1 - 5 ug/IP. Use reported in scientific literature, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000. Use reported in scientific literature (PMID 20042684), Immunohistochemistry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:50, Immunohistochemistry-Frozen 1:20-1:50, Immunoassay reported in scientific literature (PMID 26147748), Immunoblotting reported in multiple pieces of scientific literature, In vitro assay reported in multiple pieces of scientific literature, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 22411794), Proximity Ligation Assay reported in scientific literature (PMID 27595394), Tissue Culture Substratum, Ligand Activation reported in scientific literature (PMID 26147748), Immunohistochemistry Free-Floating reported in scientific literature (PMID 33242463), Chromatin Immunoprecipitation (ChIP) 1-5 ug/IP, Knockout Validated reported in scientific literature (PMID 26861754), KnockDown Validated reported in scientific literature (PMID 32772041) |
|---|---|
| Classificazione | Monoclonal |
| Target Species | Mouse, Pig, Bovine, Monkey, Rabbit |
| Alias gene | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1 |
| Isotype | IgG2b |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18741444
|
Novus Biologicals
NB100-105SS |
317.00€
25µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||||
|
18769473
|
Novus Biologicals
NB100-105 |
0.1 mL |
771.00€
0.10mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Feline, Ferret, Monkey, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Immunoblotting, In vitro assay, Gel Super Shift Assays, Proximity Ligation Assay, Tissue Culture Substratum, Ligand Activation, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated.Specifica
| Western Blot 1:500, Simple Western, Chromatin Immunoprecipitation 1 - 5 ug/IP. Use reported in scientific literature, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000. Use reported in scientific literature (PMID 20042684), Immunohistochemistry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:50, Immunohistochemistry-Frozen 1:20-1:50, Immunoassay reported in scientific literature (PMID 26147748), Immunoblotting reported in multiple pieces of scientific literature, In vitro assay reported in multiple pieces of scientific literature, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 22411794), Proximity Ligation Assay reported in scientific literature (PMID 27595394), Tissue Culture Substratum, Ligand Activation reported in scientific literature (PMID 26147748), Immunohistochemistry Free-Floating reported in scientific literature (PMID 33242463), Chromatin Immunoprecipitation (ChIP) 1-5 ug/IP, Knockout Validated reported in scientific literature (PMID 26861754), KnockDown Validated reported in scientific literature (PMID 32772041) | |
| Mouse, Pig, Bovine, Monkey, Rabbit | |
| IgG2b | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Monoclonal | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1 | |
| Protein G purified | |
| 93 kDa |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto