Learn More
UVP High Intensity Wafer Inspection Lamp
Click to view available options
Wavelength:
365 nm
Dimensione della confezione:
Pezzo
Specifica
Specifica
| Tensione | 115 V |
| Wattage | 100 W |
| Wavelength | 365 nm |
| Modello | B-100YP |
| Product Type | High Intensity Wafer Inspection Lamp |