Learn More
UVP High-Performance Uv Variable Intensity Transilluminator
Codice prodotto. 12833938
Click to view available options
Wavelength:
254 nm
Dimensione della confezione:
Pezzo
Codice prodotto. 12833938
Marca: UVP 95043002
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
UVP's high performance transilluminators feature 25W tubes and a specially constructed filter that provides the most uniform light emission possible from a tube based unit.
The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. High, medium and low intensity settings make the unit suitable for both visualisation and band cutting. An internal fan improves the tube lifetime.Dimensions: 337mm x 486mm x 143mm
Specifica
Wavelength 254 nm