Learn More
Nabertherm High Temperature Furnace LHT 08/16
Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Include:
Furnace Only
Gas Connection
Gas Connection, Over-Temp Limiter
Over-Temperature Limiter
Descrizione
- Tmax 1600°C, 1750°C, or 1800°C
- High-quality heating elements made of molybdenum disilicide offer very good protection against chemical interaction between charge and heating elements
- Adjustable air inlet opening
- Exhaust air opening in the roof
- Controller with touch operation P580 (50 programs with each 40 segments)
- Available with:
- Over-temperature limiter with adjustable cutout temperature as temperature limiter to protect the furnace and load
- Port for thermocouple in the furnace roof
- Protective gas connection to purge with non-flammable process, not gas tight
- Manual or automatic gas supply system
- Stackable saggars for loading in up to two or three levels, depending on model
Specifica
Specifica
| Tipo di unità di controllo | P580 |
| Capacità (sistema metrico) | 8 L |
| Temperatura (sistema metrico) | 1600°C |
| Tipo | Muffle Furnace with Lift Door |
| Include | Furnace Only |
| Tensione | 400 V |