missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-41285MFV500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Histone Deacetylase 2/HDAC2 Polyclonal antibody specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifica
| Histone Deacetylase 2/HDAC2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence | |
| EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human HDAC2. The immunogen is located within the last 50 amino acids of HDAC2. Amino Acid Squence: KKARIEEDKKETEDKKTD | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific, Stem Cell Markers | |
| 3066 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto