Learn More
Simport™ Scientific Histosette™ I Biopsy Processing/Embedding Cassette
Designed to hold biopsy specimens during the embedding process. Histosette™ I Biopsy Processing/Embedding Cassette are suitable to be used with some models of cassette labeling instruments.
Specifica
| Chiusura inclusa | Yes |
|---|---|
| Angolo della faccia di scrittura | 45° |
| Materiale | Acetal |
| Product Type | Biopsy Cassette |
Descrizione
- Made of high density polymer, acetal
- Keeps specimens safely submerged in liquid
- Resistant to chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely Available in 11 colors
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifica
| Yes | |
| 45° | |
| Acetal | |
| Biopsy Cassette |
For in vitro diagnostic (IVD) use only.